Learn More
Chemglass Life Sciences Chemglass pipette tip, 300 μL, standard barrier, sterile - bulk (4,800/Case)
SDP
Supplier:  Chemglass Life Sciences CLS3499D300
Encompass_Preferred
This sterile 300 μL barrier pipette tip features a fine-point, low-retention design for accurate microvolume dispensing and a hydrophobic aerosol barrier to prevent carry-over and protect pipettors.
  • Volume: 300 μL.
  • Length: standard.
  • Material: 100% polypropylene.
  • Sterilization: gamma-irradiated, sterile.
  • Certified free of RNase, DNase, human DNA, and pyrogens.
  • Hydrophobic aerosol barrier to prevent contamination.
  • Pack type: bulk packed; case quantity 4,800 per case.
  • Intended use: molecular biology and precision liquid handling applications.
Catalog No. 50-273-3213