Learn More
Chemglass Life Sciences Pipette Tips Refill for 1,000 μL Pipettors, Clear, Non-Sterile, 960/Pack
SDP
Supplier:  Chemglass Life Sciences CLS3499G100
Encompass_Preferred
Clear, non-sterile refill pipette tips for 1,000 μL volume transfer. Supplied in bulk nest format for repacking or high-throughput use, these ultra-clear, low-retention tips are compatible with most single- and multichannel pipettors and are intended for routine laboratory applications.
  • 1000 μL volume capacity.
  • 960 tips per pack, 4,800 tips per case.
  • Ultra-clear polymer resists organic solvents.
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free.
  • Low-retention, fine-point design reduces sample adhesion.
  • Universal fit for most single and multichannel pipettors.
Catalog No. 50-273-3204