Learn More
Chemglass Life Sciences Chemglass Pipette Tips - 10 μL Standard, Clear, Non-Sterile; Refill Nest, Universal Fit, 960/Pack
SDP
Supplier:  Chemglass Life Sciences CLS3499G018
Encompass_Preferred
Chemglass 10 μL standard pipette tips are clear, ultra-clear polymer refill nests designed for routine liquid handling. They provide a universal fit for most single- and multichannel pipettors and are intended for non-sterile, high-volume refill use.
  • 10 μL volume.
  • Made from ultra-clear polymer for visual clarity.
  • Non-sterile packaging suitable for general laboratory use.
  • Low-retention, fine-point design to minimize sample adhesion.
  • Certified RNase-, DNase-, human DNA-, and pyrogen-free.
  • Universal fit for most single- and multichannel pipettors.
  • Packaging: 960 per pack, 9,600 per case.
Catalog No. 50-273-3205