Learn More
Chemglass Life Sciences Pipette Tip Refills, 300 μL, Clear, Non-Sterile, Nest (960/Pack)
SDP
Supplier:  Chemglass Life Sciences CLS3499G300
Encompass_Preferred
Clear, non-sterile 300 μL pipette tip refills supplied in nest format for general laboratory pipetting and sample transfer. Packs of 960 (9,600 per case) are made from ultra-clear polymer and certified free of RNase, DNase, human DNA, and pyrogens. Low-retention, fine-point geometry and solvent compatibility reduce sample carryover and improve accuracy.
  • Volume: 300 μL.
  • Pack size: 960 per pack; 9,600 per case.
  • Material: ultra-clear polymer.
  • Sterility: non-sterile.
  • Certified RNase, DNase, human DNA, and pyrogen free.
  • Low-retention, fine-point design reduces sample adhesion.
  • Effective hydrophobic aerosol barrier minimizes carry-over.
  • Universal-fit for most single and multichannel pipettors.
Catalog No. 50-273-3201