Learn More
Chemglass Life Sciences Pipette Tips, Large Volume, 10mL, Blue, Racked (10 Racks of 25), Sterile
SDP
Supplier:  Chemglass Life Sciences CLS3499T11
Encompass_Preferred
Large volume pipette tips for use with CLS-3489E Accupet pipettors are made from high purity virgin polypropylene and are designed for use in a wide variety of pipetting applications. RNase/DNase Free. Available sterile or non-sterile in racks. Made In USA. Note: Tips can only be used with Accupet Pipettors
Catalog No. 50-009-80717