Learn More
Electron Microscopy Sciences Pipettes Volumetric 10 ml, 0.1 ml gradations
SDP
Supplier:  Electron Microscopy Sciences 7095210
Encompass_Preferred
Made from translucent polypropylene. Resistant to breakage. Blue graduations, ring mark and inscriptions. Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 degree C. NOTE: Exposure to temperatures above 60 degree C may cause volume changes.
Capacity: 10 ml
Gradations: 0.1 ml
Catalog No. 50-189-9380