Learn More
Beckman Coulter PIPETTING HEAD MC96 5-1200UL
Supplier:  Beckman Coulter B87589
Encompass_Preferred
  • 96 multichannel layout.
  • Volume range of 5-1200 µL.
  • Compatible with Biomek i-Series automated workstations.
  • Enables automated liquid handling.
  • Utilizes disposable tips to prevent cross-contamination.
  • Ideal for high-throughput pipetting applications.
  • Supports sample transfer and liquid mixing functions.
  • Delivers precise and reproducible results.
Catalog No. NC1890821
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.