Learn More
Thomas Scientific Siliconized Pipettor Tips 1-200 uL, 500/pk
SDP
Supplier:  Thomas Scientific 7738E05
Encompass_Preferred
SILCONIZED TIPS minimize liquid retention of viscous or proteinaceous samples.Precise and complete sample deliveryPrevents protein or nucleic acid adherenceUniversal fit provides absolute sealIdeal for use in DNA procedures. Low adhesion surface assures complete transfer of samples. Specially formulated siliconized polypropylene helps to eliminate sample residue and cross-contamination. Designed for use with PIPETMAN®, Eppendorf, Finnpipette and Socorex pipettors. PIPETMAN® is a registered trademark of Gilson, Inc.
Catalog No. NC3342878