Learn More
Selleck Chemical LLC Pipobroman S4447-25mg
SDP
Supplier:  Selleck Chemical LLC S444725MG
Encompass_Preferred
Pipobroman (PB Vercyte) a neutral amide of piperazine acts as an alkylating agent with antiproliferative activity
Catalog No. 50-311-1182