Learn More
Aviva Systems Biology Corp PIR Antibody - C-terminal region (ARP33665_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33665_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PIR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PIR. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55110