Learn More
Medchemexpress LLC Pirarubicin (hydrochloride) | 95343-20-7 | 99.1% | 664.10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13725A100MG
Encompass_Preferred
Pirarubicin Hydrochloride is an anthracycline antibiotic that acts as a topoisomerase II inhibitor and is widely used for the treatment of various cancers, particularly solid tumors. This compound induces autophagy in bladder cancer cells in a dose-dependent manner and also induces apoptosis.
  • Acts as a topoisomerase II inhibitor
  • Widely used for the treatment of various cancers
  • Induces autophagy in bladder cancer cells
  • Induces apoptosis by inhibiting mTOR/p70S6K/4E-BP1 pathway
Catalog No. 50-003-49620