Learn More
Apexbio Technology LLC Pirenzepine dihydrochloride 29868-97-1 100mg
SDP
Supplier:  Apexbio Technology LLC B6582100
Encompass_Preferred
Pirenzepine dihydrochloride (CAS 29868-97-1) is a selective antagonist of the muscarinic acetylcholine M1 receptor By inhibiting M1 receptor-mediated signaling it effectively suppresses gastric acid secretion and alleviates smooth muscle contractions These properties have supported its use in studies related to peptic ulcer disease where modulation of gastric secretory activity is relevant Additionally pirenzepine dihydrochloride has been investigated for its capacity to inhibit cancer cell proliferation making it valuable for research into tumor biology and potential therapeutic strategies targeting the cholinergic system
Catalog No. 50-000-92102