Learn More
Apexbio Technology LLC Pirfenidone, 1g Cas# 53179-13-8
SDP
Supplier:  Apexbio Technology LLC B22881000
Encompass_Preferred
Pirfenidone is an inhibitor for TGF- beta production and TGF- Beta stimulated collagen production, reduces production of TNF- alpha and IL-1B, and also has anti-fibrotic and anti-inflammatory properties. Phase 3. 53179-13-8. MFCD00866047 Pirfenidone,an oral antifibrotic agent, has a broad-spectrum of antifibroticand anti-inflammatory effects. Pirfenidonehas beneficial effects for the treatment of certain fibrotic diseases, and is under clinical trials in patients with idiopathic pulmonary fibrosis. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8697