Learn More
Apexbio Technology LLC Pirfenidone 53179-13-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B22885.1
Encompass_Preferred
Pirfenidone (CAS 53179-13-8) is a small molecule antifibrotic agent exhibiting anti-inflammatory properties Mechanistically pirfenidone reduces levels of pro-inflammatory mediators notably TNF- via translational regulation independent of MAPK pathway activation It suppresses TGF- -mediated signaling and extracellular matrix protein production contributing to decreased fibrotic responses in cellular and animal models In vivo pirfenidone administration mitigates fibrosis and inflammatory cytokine expression in tissues such as lung liver and kidney Clinically pirfenidone is investigated primarily for idiopathic pulmonary fibrosis demonstrating reductions in fibrosis progression and improved functional parameters
Catalog No. 50-000-63687