Learn More
Medchemexpress LLC Pirlimycin hydrochloride | 78822-40-9 | MFCD27952783 | 99.1% | 447.42 | C17H32Cl2N2O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY106597A5MG
Encompass_Preferred
Pirlimycin hydrochloride is the hydrochloride salt of pirlimycin, a lincosamide-class antibiotic provided as a research-grade reagent. It is primarily active against Gram-positive bacteria and is supplied as a purified, stable salt for laboratory applications.
Catalog No. 50-002-41023