Learn More
Medchemexpress LLC Pirlindole | 60762-57-4 | MFCD00447212 | 98.9% | 226.32 g/mol | C15H18N2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1006791MG
Encompass_Preferred
Pirlindole is a selective, reversible monoamine oxidase A (MAO-A) inhibitor with reported antiviral activity against enterovirus D68 and coxsackievirus B3. Supplied as a high-purity research chemical, it is intended for biochemical and virology studies and analytical applications.
  • Selective, reversible MAO-A inhibition.
  • Reported antiviral activity against enterovirus D68 and coxsackievirus B3.
  • High chemical purity suitable for research use.
  • Intended for biochemical and virology assays.
  • Recommended sealed storage at -20°C; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-13717