Learn More
Medchemexpress LLC Pirlindole | 60762-57-4 | MFCD00447212 | 98.9% | 226.32 g/mol | C15H18N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10067910MG
Encompass_Preferred
Pirlindole is a selective, reversible monoamine oxidase A (MAO-A) inhibitor used as a research reagent in biochemical and virology studies. It has reported antiviral activity against enterovirus-D68 and coxsackievirus B3 (CV-B3). The compound is provided as a high-purity research grade material suitable for enzyme inhibition assays, antiviral screening, and pathway investigation.
  • Selective and reversible MAO-A inhibition suitable for enzyme assays.
  • Reported antiviral activity against enterovirus-D68 and coxsackievirus B3.
  • High purity for reliable experimental reproducibility.
  • Well-characterized molecular properties (C15H18N2, 226.32 g/mol).
  • Available in small-scale quantities for screening and validation studies.
Catalog No. 50-002-13834