Learn More
Medchemexpress LLC Piromelatine | 946846-83-9 | MFCD30146157 | 99.5% | 312.32 g/mol | C17H16N2O4 | 5 MG

Supplier: Medchemexpress LLC HY1052855MG
Piromelatine is a multimodal neuropharmacological research compound acting as a melatonin MT1/MT2 receptor agonist, serotonin 5-HT1A/5-HT1D agonist, and 5-HT2B antagonist. It has been investigated for sleep-promoting, analgesic, anxiolytic, antidepressant, and potential neuroprotective effects. This material is supplied for laboratory research use only.
- Acts as a melatonin MT1/MT2 receptor agonist.
- Modulates serotonin receptors (5-HT1A/5-HT1D agonism, 5-HT2B antagonism).
- Reported to inhibit pain-related P2X3, TRPV1, and Nav1.7 channels.
- Molecular formula C17H16N2O4; molecular weight 312.32 g/mol.
- Recommended storage: 4°C for solid; protect from light; in solvent store at -80°C or -20°C.
- For research use only; not for human or clinical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.