Learn More
Apexbio Technology LLC Piromidic Acid 19562-30-2 50mg
SDP
Supplier:  Apexbio Technology LLC A850250
Encompass_Preferred
Piromidic acid (CAS 19562-30-2) is a small-molecule inhibitor targeting bacterial DNA gyrase It is designed to inhibit DNA gyrase activity thereby disrupting bacterial DNA replication and cell proliferation Piromidic acid exerts its biological activity primarily through inhibition of bacterial DNA gyrase In in vitro studies piromidic acid demonstrates antibacterial activity with reported IC50 values for DNA gyrase inhibition in the low micromolar range dependent on the bacterial strain and assay conditions Based on these pharmacological properties piromidic acid holds research potential in investigating bacterial susceptibility resistance mechanisms and DNA gyrase function especially in the context of pathogens responsible for intestinal biliary and urinary tract infections
Catalog No. 50-000-95757