Learn More
Cayman Chemical PIroxIcam-d3 1mg
SDP
Supplier:  Cayman Chemical 332481 MG
Encompass_Preferred
An internal standard for the quantification of piroxicam by GC- or LC-MS
Catalog No. 50-294-9202