Learn More
Cayman Chemical PIrtobrutInIb 10mg
SDP
Supplier:  Cayman Chemical 3677510 MG
Encompass_Preferred
A BTK inhibitor (IC50s = 10-100 and C481S, respectively); selective for BTK over EGFR (IC50 = 1-10 µM); decreases the number of Ki67-positive cells in MEC-1 CLL mouse xenograft models using MEC-1 cells that express either wild-type BTK or BTKC481S
Catalog No. 50-291-3321