Learn More
Selleck Chemical LLC Pirtobrutinib S9825-100MG
SDP
Supplier:  Selleck Chemical LLC S9825100MG
Encompass_Preferred
Pirtobrutinib (LOXO-305 LY 3527727 RXC-005) is a highly selective non-covalent next generation BTK inhibitor with an IC50 of 5 69 nM in WT BTK HEK cells Pirtobrutinib shows more than 300-fold selective for BTK over 98% of 370 other kinases
Catalog No. 50-000-24835