Learn More
Medchemexpress LLC Pisiferic acid | 67494-15-9 | MFCD01708794 | 99.2% | 316.43 | C20H28O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW3548515MG
Encompass_Preferred
Pisiferic acid is an antibacterial research compound (CAS 67494-15-9) that exhibits inhibitory activity against Gram-negative and Gram-positive bacteria such as Proteus vulgaris, Staphylococcus aureus, and Bacillus subtilis. Supplied as a white to off-white solid, it is intended for in vitro antibacterial assays and microbiology research.
  • Antibacterial activity against Gram-negative and Gram-positive bacteria.
  • High purity (≈99.2%) suitable for analytical and biological studies.
  • Solid, white to off-white appearance for straightforward handling.
  • Stable when stored as powder at -20°C and in solution at -80°C for extended periods.
  • Available in small research quantities, including 5 mg packages.
Catalog No. 50-002-85187