Learn More
Bioworld PISUM SATIVUMGARDEN PEA)
Supplier:  Bioworld 215100092
Encompass
PISUM SATIVUM (GARDEN PEA) - LECTIN: Pisum sativum (Garden pea) Lectin isolated from Pisum sativum (Garden pea) Specifications: Primary Sugar Specificity: Glucose/Mannose Preferential binding activity: alpha-linked mannose (requir...
Catalog No. 50-198-741