Learn More
American Radiolabeled Chemicals Inc PITAVASTATIN 3-3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 194650UC
Encompass_Preferred
Pitavastatin-3-3H 50uCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 421.46, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-982
Add to Cart
Request a Quote