Learn More
Medchemexpress LLC Pitavastatin impurity 12 | 185105-98-0 | 366.39 | 10 G
SDP
Supplier:  Medchemexpress LLC HY2131510G
Encompass_Preferred
Pitavastatin impurity 12 is an impurity of Pitavastatin (HY-B0144A). It is for research use only.
  • Not for sale to patients
  • Solid appearance
  • Store at room temperature for 3 years, or in solvent at -80°C for 2 years or -20°C for 1 year
Catalog No. 50-004-00714