Learn More
Cayman Chemical PItavastatIn lacton-d4 500ug
SDP
Supplier:  Cayman Chemical 34696500 UG
Encompass_Preferred
An internal standard for the quantification of pitavastatin lactone by GC- or LC-MS
Catalog No. 50-291-1364