Learn More
Sklar Instruments Pitcher/Beaker, Sklar, Stainless Steel. This product is 6-7/8 in. x 8-7/8 in. with a volume of 4000ml. It features a closed handle. EA/1.
SDP
Supplier:  Sklar Instruments 101181
Encompass_Preferred
Pitcher/Beaker, Sklar, Stainless Steel. Sklar's Stainless Steel Beaker may be used for a variety of things, including use in the laboratory, or holding sterilized instruments in solution until their use in specific procedures. This product is 6-7/8 in. x 8-7/8 in. with a volume of 4000ml. It features a closed handle.
Catalog No. 50-143-6944