Learn More
Medchemexpress LLC Pitofenone hydrochloride | 1248-42-6 | 99.9% | 403.90 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY110389100MG
Encompass_Preferred
Pitofenone hydrochloride is a spasmolytic compound that inhibits acetylcholinesterase (AChE) activity. It exhibits anticonvulsant activity when combined with diclofenac and fenpivirine.
  • Inhibits acetylcholinesterase (AChE) activity.
  • Exhibits anticonvulsant activity.
  • Suitable for neurological disease research.
Catalog No. 50-003-44051