Learn More
TARGETMOL CHEMICALS INC PITOLISANT HYDROCHLORIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T717625MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pitolisant hydrochloride (Ciproxidine) is a potent and selective inverse agonist of the histamine H3 receptor(Ki 0.16 nM). purity: 99%
Catalog No. 50-257-6340