Learn More
ABclonal Technology PITPNA Rabbit pAb
SDP
Supplier:  ABclonal Technology A129661000UL
Encompass_Preferred
This gene encodes a member of a family of lipid-binding proteins that transfer molecules of phosphatidylinositol or phosphatidylcholine between membrane surfaces. The protein is implicated in phospholipase C signaling and in the production of phosphatidylinositol 3,4,5-trisphosphate (PIP3) by phosphoinositide-3-kinase.
Catalog No. 50-001-14643