Learn More
TARGETMOL CHEMICALS INC PITSTOP 2 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2635710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pitstop2 is a selective inhibitor of amphiphysin association of clathrin terminal domain with an IC50 value of 12 uM. Pitstop2 acutely interferes with receptor-mediated endocytosis entry of HIV and synaptic vesicle recycling. purity: 99%
Catalog No. 50-257-7075