Learn More
Medchemexpress LLC Pitstop 2 25mg | 1419320-73-2 | 473.36 g/mol | C20H13BrN2O3S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11560425MG
Encompass_Preferred
Pitstop 2 is a small-molecule inhibitor of clathrin-mediated endocytosis that binds the terminal domain of clathrin. It is used in cell biology and cancer-research studies to probe endocytic pathways and assess clathrin-dependent uptake.
  • Inhibits clathrin-mediated endocytosis by targeting the clathrin terminal domain.
  • Suitable for mechanistic studies of cellular uptake and endocytic trafficking.
  • Soluble in DMSO (≈50 mg/mL); handle DMSO carefully due to hygroscopicity.
  • Provided at high purity (~98.4%) for research applications.
  • Available in small research pack sizes appropriate for in vitro experiments.
Catalog No. 50-002-29678