Learn More
Selleck Chemical LLC Pitstop 2 5mg
SDP
Supplier:  Selleck Chemical LLC S96705MG
Encompass_Preferred
Pitstop 2 is an inhibitor of the interaction of amphiphysin with the amino terminal domain of clathrin that inhibits both clathrin-dependent endocytosis (CDE) and clathrin independent endocytosis (CIE). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3489