Learn More
Medchemexpress LLC Pivalate-d3 (sodium) | 39230-37-0 | 85.05 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW006398S1G
Encompass_Preferred
Pivalate-d3 (sodium) is the deuterium labeled acetic acid. It is a carboxylic acid and short-chain fatty acid (SCFAs). This compound activates AMPK, increases ROS, cleaved caspase 9, PPARα, and downregulates SREBP-1c and ChREBP expression. It exhibits antifungal activity against *Saccharomyces cerevisiae* W303-1A, regulates energy metabolism, and has anticancer activity against gastric cancer. It can be used in research for gastric cancer, ulcerative colitis, hepatic steatosis, and pain.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antifungal activity
  • Regulates energy metabolism
  • Has anticancer activity against gastric cancer
  • Induces writhing reaction and ulcerative colitis
Catalog No. 50-004-46355