Learn More
Medchemexpress LLC Pivalyloxymethyl butyrate | 122110-53-6 | >=95.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY120508100MG
Encompass_Preferred
Pivalyloxymethyl butyrate (AN-9) is a liquid histone deacetylase (HDAC) inhibitor used in research applications such as epigenetic modulation and preclinical oncology studies. It is typically supplied for laboratory use and may be prepared as solutions in DMSO for in vitro assays.
  • Small-molecule HDAC inhibitor that modulates histone acetylation and gene expression.
  • Liquid form suitable for solution preparation and in vitro assays.
  • Typical purity approximately 95% by assay.
  • Molecular weight 202.25 g/mol and formula C10H18O4.
  • Commonly used in epigenetic research and preclinical cancer studies.
Catalog No. 50-002-32900