Learn More
Selleck Chemical LLC Pivmecillinam hydrochloride S5473-1g
SDP
Supplier:  Selleck Chemical LLC S54731G
Encompass_Preferred
Pivmecillinam is a -lactam antibiotic and a prodrug of mecillinam It has selective activity against Gram-negative bacteria and is used primarily in the treatment of lower urinary tract infections
Catalog No. 50-311-0285