Learn More
Medchemexpress LLC Pivmecillinam hydrochloride | 32887-03-9 | 98.54% or 99.19% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0810A100MG
Encompass_Preferred
Pivmecillinam hydrochloride (FL-1039 hydrochloride) is an orally active proagent of mecillinam, an extended-spectrum penicillin antibiotic.
  • Orally active proagent
  • Extended-spectrum penicillin antibiotic
  • Effective in inhibiting urinary tract infections caused by multidrug-resistant E. coli in a mouse model
Catalog No. 50-004-09272