Learn More
Selleck Chemical LLC Pixantrone Maleate S5059-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S505910MM/1ML
Encompass_Preferred
Pixantrone (BBR-2778) is a novel aza-anthracenedione compound with antitumor activity It is a weak topoisomerase II inhibitor and forms stable DNA adducts through alkylation with specificity for DNA hypermethylated sites
Catalog No. 50-311-1965