Learn More
Cayman Chemical PJ-34hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 144405 MG
Encompass_Preferred
An inhibitor of PARPs which can be used in cells or in animals; binds and inhibits the PARP TNKS1 (IC50 = 1 μM); inhibits MMP-2 when used at higher concentrations (IC50 = 56 μM)
Catalog No. 50-297-0925