Learn More
TARGETMOL CHEMICALS INC PJ34 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T619750MG
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PJ34 HCl is the hydrochloride salt of PJ34 which is a PARP inhibitor with EC50 of 20 nM and is equally potent to PARP1/2. purity: 99%
Catalog No. 50-257-9493