Learn More
Ace Glass, Inc. PK3 7/10 PTFE SLEEVE PK3 7/10 PTFE SLEEVE
SDP
Supplier:  Ace Glass, Inc. 764202
Encompass
7/10 ground joint standard taper PTFE sleeve for greaseless connections. NOT intended for cementing. Wall thickness is 0.13mm (0.005¿). Pack of 3.
Catalog No. 50-975-198