Learn More
Sino Biological Peptide Substrates: PKA Substrate
Supplier:  Sino Biological C0158
Encompass_Preferred
The PKA sub peptide sequence (CGRTGRRNSI-amide) is based on the cAMP-dependent protein kinase inhibitor alpha (amino acid 15-23).
Catalog No. 50-256-9660