Learn More
Enzo Life Sciences Protein kinase C (20-28) (myristoylated) (0.5mg)
SDP
Supplier:  Enzo Life Sciences BMLP2050500
Encompass_Preferred
Pseudosubstrate sequence from PKCα and ß. Highly specific inhibitor, which is N-terminal myristoylated to allow membrane permeability. IC50=8 µM for TPA activation of MARCKS phosphorylation in fibroblast primary cultures, and 98% inhibition at 100 µM. Partially reverses multidrug resistance in human breast cancer cells. Alternative name: PKC (20-28) (myristoylated). Purity: ≥98% (HPLC). Formulation: TFA salt. Lyophilized. Solubility: Soluble in water or in 20mM TRIS-HCl, pH 7.5. Long Term Storage: -20°C.
Catalog No. 50-201-1282