Learn More
Medchemexpress LLC PKC-theta inhibitor 2 | 736048-65-0 | 99.9% | 454.45 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1126811MG
Encompass_Preferred
PKC-theta inhibitor 2 is a potent and selective PKC-θ inhibitor with an IC50 value of 18 nM. It is a 2,4-diamino-5-nitropyrimidine compound with 2-position substituted by o-trifluoromethoxy. It is intended for research use only.
  • Potent and selective PKC-θ inhibitor
  • IC50 value of 18 nM
  • 2,4-diamino-5-nitropyrimidine compound
  • 2-position substituted by o-trifluoromethoxy
Catalog No. 50-004-36994