Learn More
Medchemexpress LLC HY-112681 5mg Medchemexpress, PKC-theta inhibitor CAS:736048-65-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126815MG
Encompass_Preferred
Medchemexpress, HY-112681 5mg PKC-theta inhibitor CAS:736048-65-0 PKC-theta inhibitor is a selective PKC-θinhibitor, with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9139