Learn More
Medchemexpress LLC PKG inhibitor peptide | 82801-73-8 | 99.8% | 943.11 | 5 MG

Supplier: Medchemexpress LLC HYP12925MG
PKG inhibitor peptide is an ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG) with a Ki of 86 μM. It is intended for research use only.
- ATP-competitive inhibitor of cGMP-dependent protein kinase (PKG)
- Ki of 86 μM for PKG
- Solid appearance, white to off-white color
- Soluble in DMSO and H2O at ≥ 100 mg/mL
- Recommended storage as powder at -80°C for 2 years or -20°C for 1 year (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.