Learn More
Medchemexpress LLC Pkg substrate | 81187-14-6 | 99.0% | 902.01 | 10 MG

Supplier: Medchemexpress LLC HYP156110MG
PKG Substrate is a selective substrate for cGMP-dependent protein kinase (PKG). This product appears as a solid, white to off-white substance with the sequence Arg-Lys-Arg-Ser-Arg-Ala-Glu (RKRSRAE) and is intended for research use only.
- Selective for cGMP-dependent protein kinase (PKG)
- Solid, white to off-white appearance
- Sequence: Arg-Lys-Arg-Ser-Arg-Ala-Glu (RKRSRAE)
- Powder storage: -80°C for 2 years or -20°C for 1 year in sealed storage, away from moisture and light, under nitrogen
- In solvent storage: -80°C for 6 months or -20°C for 1 month in sealed storage, away from moisture and light, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.