Learn More
Medchemexpress LLC PKI-166 | 330.38 g/mol | C20H18N4O | 100MG
SDP
Supplier:  Medchemexpress LLC HY117155100MG
Encompass_Preferred
PKI-166 is a potent selective and orally bioavailable EGFR tyrosine kinase inhibitor with an IC50 of 0 7 nM[1]
Catalog No. 50-002-29407