Learn More
Enzo Life Sciences PKI (6-22) amide (1mg). CAS: 121932-06-7
SDP
Supplier:  Enzo Life Sciences BMLP2040001
Encompass_Preferred
Competitive inhibitor of PKA (Ki =1.7 nM). Alternative name: cAMP dependent protein kinase inhibitor (6 22) amide. Purity: ≥97% (HPLC). Formulation: Lyophilized. Trifluoroacetate salt. Solubility: Soluble in water (10 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1281